EMAIL THIS PAGE TO A FRIEND
Bioorganic & medicinal chemistry letters
Novel spiroanellated 1,2,4-trioxanes with high in vitro antimalarial activities.
PMID 15664819
Abstract
A remarkable increase in antimalarial in vitro activity was achieved by integration of spiroadamantane motifs in 6-alkylidene 1,2,4-trioxanes 3a-h via diastereoselective photooxygenation of allylic alcohols and subsequent BF(3)-catalyzed peroxyacetalization with adamantanone to give the active compounds 3e-h.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
T81108
1,3,5-Trioxane, ≥99%
C3H6O3