EMAIL THIS PAGE TO A FRIEND
Bioorganic & medicinal chemistry letters
Crystallographic determination of stereochemistry of biologically active 2'',3''-dibromo-7-epi-10-deacetylcephalomannine.
PMID 15664868
Abstract
The stereochemistry at C2'' and C3'' of two diastereomers of 2'',3''-dibromo-7-epi-10-deacetylcephalomannine (6 and 7), which were synthesized by reacting 7-epi-10-deacetylcephalomannine (5) with bromine, were assigned unambiguously based on crystallographic studies of 6. The X-ray crystallographic analysis shows that 6 adopts an absolute configuration of (2''S,3''R), and 7 can be assigned as (2''R,3''S) configuration. The side-chain conformation of 6 was revealed to be different with the known hydrophobic collapse and the apolar conformations, as found in solid state and in solution. However, most side-chain torsion angles of 6 were found to be very similar to those of tubulin-bound T-shaped conformation (T-Taxol). Both 6 and 7 showed strong in vitro paclitaxel-like activity.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
C4991
Cephalomannine, ≥97% (HPLC), solid
C45H53NO14