EMAIL THIS PAGE TO A FRIEND
Zeitschrift fur Naturforschung. C, Journal of biosciences
Induction of tropane alkaloid formation in transformed root cultures of Brugmansia suaveolens (Solanaceae).
PMID 15666547
Abstract
Hairy root cultures of Brugmansia suaveolens were set up by infection of root tips with Agrobacterium rhizogenes. The successful transformation was confirmed by analysing rolC and virC genes using polymerase chain reaction (PCR). Hairy root cultures were employed to study the formation of tropane alkaloids, such as hyoscyamine. The transformed cultures were incubated with potential elicitors, such as methyljasmonate, quercetin and salicylic acid in order to stimulate the biosynthesis of tropane alkaloids. Profile and amounts of tropane alkaloids were analysed using capillary GLC-MS. At least 18 different tropane alkaloids could be identified. Treatment of the cultures with 200 microM methyljasmonate increased the alkaloid accumulation 25-fold up to a level of 1 mg/g fresh weight as compared to untreated controls. Quercetin enhanced the alkaloid production 10 fold (0.4 mg/g fresh weight) within 24 h. In contrast 100 microM salicylic acid decreased alkaloids to a level of 1 microg/g fresh weight.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
93550
Tropine, ≥97.0% (NT)
C8H15NO