EMAIL THIS PAGE TO A FRIEND
Bulletin of environmental contamination and toxicology
Distribution and dissipation of phenthoate insecticide following aerial application.
PMID 15669725
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
31611
Phenthoate, PESTANAL®, analytical standard
C12H17O4PS2