EMAIL THIS PAGE TO A FRIEND
Dalton transactions (Cambridge, England : 2003)
Synthesis, characterization and coordination chemistry of a macrocyclic ligand containing arsenic donors.
PMID 15672188
Abstract
The preparation and characterization of the macrocyclic diamido-diarsine ligand [As2N2]Li2(1,4-dioxane) (1) (where As2N2 = PhAs(CH2SiMe2NSiMe2CH2)2AsPh) and a series of early transition metal complexes are presented. The complexes [As2N2]MCl2 (M = Zr, 2; Ti, 4) and the complex ([As2N2]Y)2(mu-Cl)2 (5) can be prepared by reaction of 1 with the corresponding THF adduct of the metal halide. The iodide derivative of 2, [As2N2]ZrI2 (3) can be prepared by reaction with iodotrimethylsilane. The lithium complex 1 displays a very long lithium-arsenic bond distance of 3.162(10) A, and the yttrium complex 5 is the first known complex containing a yttrium-arsenic bond. Reduction of 2, 3 or 4 using C8K or activated magnesium decomposed the complexes in such a manner that the ligand was separated from the metal centre. Indirect evidence suggests this may be due to reduction of arsenic in the ligand in preference to the metal.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
234990
1,3-Bis(chloromethyl)-1,1,3,3-tetramethyldisilazane, 95%
C6H17Cl2NSi2