EMAIL THIS PAGE TO A FRIEND
Antimicrobial agents and chemotherapy
Antifungal susceptibilities of clinical isolates of Candida species, Cryptococcus neoformans, and Aspergillus species from Taiwan: surveillance of multicenter antimicrobial resistance in Taiwan program data from 2003.
PMID 15673726
Abstract
The susceptibilities of nonduplicate isolates to six antifungal agents were determined for 391 blood isolates of seven Candida species, 70 clinical isolates (from blood or cerebrospinal fluid) of Cryptococcus neoformans, and 96 clinical isolates of four Aspergillus species, which were collected in seven different hospitals in Taiwan (as part of the 2003 program of the study group Surveillance of Multicenter Antimicrobial Resistance in Taiwan). All isolates of Candida species other than C. glabrata and C. krusei were susceptible to fluconazole. Among the 59 C. glabrata isolates, 16 (27%) were not susceptible to fluconazole, and all were dose-dependently susceptible or resistant to itraconazole. For three (5.1%) C. glabrata isolates, voriconazole MICs were 2 to 4 microg/ml, and for all other Candida species isolates, voriconazole MICs were /=2 microg/ml were 100% (3 isolates) for C. krusei, 11% (23 of 207 isolates) for Candida albicans, 3.0% (2 of 67 isolates) for Candida tropicalis, 20% (12 of 59 isolates) for C. glabrata, and 0% for both Candida parapsilosis and Candida lusitaniae. For three (4%) Cryptococcus neoformans isolates, fluconazole MICs were >/=16 microg/ml, and two (3%) isolates were not inhibited by 1 mug of amphotericin B/ml. For four (4.2%) of the Aspergillus isolates, itraconazole MICs were 8 microg/ml. Aspergillus flavus was less susceptible to amphotericin B, with the MICs at which 50% (1 microg/ml) and 90% (2 microg/ml) nsrsid417869\delrsid7301351 of isolates were inhibited being twofold greater than those for Aspergillus fumigatus and Aspergillus niger. All Aspergillus isolates were inhibited by
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
UC280
Ketoconazole
C26H28Cl2N4O4