EMAIL THIS PAGE TO A FRIEND
Journal of AOAC International
Liquid chromatographic method to determine narasin in feedingstuffs and premixtures: development, validation, and interlaboratory study.
PMID 15675437
Abstract
A reversed-phase liquid chromatography (LC) method for narasin in feedingstuffs and premixtures was developed, validated, and interlaboratory studied. The extraction solvent was methanol-K2HPO4 solution (9 + 1, v/v). Narasin was detected at 600 nm after post-column derivatization with dimethylamino-benzaldehyde. Recovery was >90%. The repeatability (RSDr) in feed (20-140 mg/kg) ranged between 1.2 and 10.5%; the within-laboratory reproducibility (RSD(R)) ranged between 2.2 and 4.9%. The limit of determination was <20 mg/kg. Other feed additives did not interfere in the assay. The method showed ruggedness against changes in the composition of extraction solvent, eluent, and conditions for post-column reactions. In an interlaboratory study, 5 broiler feeds (4 positive, 1 blank) and 1 premixture were analyzed by 13 laboratories. The RSDr of the feedingstuffs (20-120 mg/kg) varied between 2.17 and 7.57%. The HORRAT ranged between 0.77 and 0.88, with recoveries between 82 and 104%. One laboratory detected small signals in the blank sample, calculated as 0.6 and 2.8 mg/kg. For the premixture, acceptable results for reproducibility could only be obtained after modification of the method: the RSDr was 4.42% and the HORRAT was 1.56 (12 laboratories).
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
N1271
Narasin from Streptomyces auriofaciens, ≥98% (HPLC)
C43H72O11