EMAIL THIS PAGE TO A FRIEND
Journal of agricultural and food chemistry
Carbohydrate accumulation in leaves of plants treated with the herbicide chlorsulfuron or imazethapyr is due to a decrease in sink strength.
PMID 15675810
Abstract
Herbicides that inhibit branched chain amino acid biosynthesis produce a rapid carbohydrate increase in leaves of treated plants. The relationship between these processes is not known nor is the importance of carbohydrate accumulation in the growth inhibition caused by these herbicides. This work analyzes carbohydrate concentration in sources and sinks after herbicide treatments in pea (Pisum sativum L.), as well as photosynthetic carbon assimilation, using two classes of chemicals, chlorsulfuron and imazethapyr, applied to roots or leaves. The most remarkable result was that, in addition to carbohydrate accumulation in leaves, accumulation of sucrose and/or starch in roots was detected. This pattern of carbohydrate accumulation was similar for both herbicides and independent of whether the herbicides were applied to leaves or roots. This indicates that root growth inhibition was not caused by sugar starvation in sinks. Nevertheless, the results are consistent with a decrease in sink strength, leading to the inhibition of photoassimilate translocation.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
34322
Chlorosulfuron, PESTANAL®, analytical standard
C12H12ClN5O4S