EMAIL THIS PAGE TO A FRIEND
American journal of clinical dermatology
The role of dimethylaminoethanol in cosmetic dermatology.
PMID 15675889
Abstract
Skincare formulations for the improvement of aging skin are increasingly important consumer products. Here, we review available data on one such agent - 2-dimethylaminoethanol (DMAE) or deanol - that has recently been evaluated in a placebo-controlled trial. DMAE is an analog of the B vitamin choline and is a precursor of acetylcholine. Although the role of acetylcholine as a neurotransmitter is well known, growing evidence points to acetylcholine as a ubiquitous cytokine-like molecule that regulates basic cellular processes such as proliferation, differentiation, locomotion, and secretion in a paracrine and autocrine fashion. Indeed, this modulatory role may contribute to the cutaneous activity of DMAE. In a randomized clinical study, 3% DMAE facial gel applied daily for 16 weeks has been shown to be safe and efficacious (p < 0.05) in the mitigation of forehead lines and periorbital fine wrinkles, and in improving lip shape and fullness and the overall appearance of aging skin. These effects did not regress during a 2-week cessation of application. Beneficial trends (p > 0.05 but
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
391263
2-Dimethylaminoethanol, purified by redistillation, ≥99.5%
C4H11NO