EMAIL THIS PAGE TO A FRIEND
Clinical chemistry
Abrupt changes in the concentrations of five prostatic tumor-associated markers in serum after acute myocardial infarction.
PMID 1568341
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
M47353
mono-Methyl glutarate, 95%
C6H10O4