EMAIL THIS PAGE TO A FRIEND
Molecular and cellular biology
Chromatin architecture near a potential 3' end of the igh locus involves modular regulation of histone modifications during B-Cell development and in vivo occupancy at CTCF sites.
PMID 15684400