EMAIL THIS PAGE TO A FRIEND
Biotechnology letters
High-yield conversion of (R)-2-octanol from the corresponding racemate by stereoinversion using Candida rugosa.
PMID 15685415
Abstract
Whole cells of Candida rugosa catalyzed the conversion of (R)-2-octanol from the corresponding racemate with the optical purity of 97% e.e. and yield of 92% in 10 h. The product was formed through a stereoinversion involving enantioselective oxidation and asymmetric reduction with 2-octanone as the intermediate.