EMAIL THIS PAGE TO A FRIEND
Bioorganic & medicinal chemistry letters
Halogenated and isosteric cytisine derivatives with increased affinity and functional activity at nicotinic acetylcholine receptors.
PMID 15686946
Abstract
A series of pyridone ring-modified derivatives of (7R,9S)-(-)-cytisine were evaluated for affinity and functional activity at neuromuscular alpha1beta1gammadelta, ganglionic alpha3beta4, and central neuronal alpha4beta2 subtypes of nicotinic receptors. Halogenation at the 3-position improved affinity and functional activity, while substitution at the 5-position led to modest decreases in both, and disubstitution led to near abolition of functional activities and could be correlated with the electron-withdrawing ability of the halogen. Subtype selectivities of the halogenated derivatives were altered relative to cytisine in a substitution-dependent manner. Caulophylline methiodide was less potent than cytisine, but retained significant activity. Thiocytisine was relatively weak in potency and efficacy, but was significantly selective for the alpha4beta2 subtype.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
C2899
Cytisine, ≥99%, powder
C11H14N2O