EMAIL THIS PAGE TO A FRIEND
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences
Determination of bromide in whole blood and urine from humans using gas chromatography-mass spectrometry.
PMID 15687004
Abstract
We devised a sensitive and simple method for determination of bromide in whole blood and urine from humans using gas chromatography-mass spectrometry. Bromide was alkylated with pentafluorobenzyl p-toluenesulphonate in the mixture of acetone and phosphate buffer (pH 6.8). The derivative obtained was analyzed using gas chromatography-mass spectrometry with the positive-ion EI mode. The lower limit of detection for the compound was 1 mg/l. The calibration curve for bromide was linear over the concentration range from 2 to 100 mg/l. With use of this method, levels of bromide in whole blood and urine were determined in cases of poisoning by inhaled brominated hydrocarbons.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
P5360
2,3,4,5,6-Pentafluorobenzoic acid, 99%
C7HF5O2