EMAIL THIS PAGE TO A FRIEND
International journal of cancer
Inhibition of malate-aspartate shuttle by the antitumor drug L-glutamic acid gamma-monohydroxamate in L1210 leukemia cells.
PMID 1568800
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
G2253
L-Glutamic acid γ-monohydroxamate
C5H10N2O4