EMAIL THIS PAGE TO A FRIEND
Dental materials journal
Durability of FTLA treatment as a medicament for dentin hypersensitivity--abrasion resistance and profiles of fluoride release.
PMID 15688723
Abstract
The purpose of this study was to evaluate the durability of tubules occluded with FTLA treatment by toothbrush abrasion test on the applied surface and by measuring fluoride release from the FTLA components. Dentin specimens with simulated hypersensitive surfaces were treated with APF containing tannic acid. After which, the specimens received lanthanum-chloride-with-powdered-fluoroapatite-glass-ceramics treatment. The specimens were subjected to toothbrush abrasion test up to 6,000 strokes. SEM observation revealed that dentinal tubules of the FTLA treated specimens were completely occluded with fine deposits even after toothbrush abrasion of 6,000 strokes. EPMA analysis revealed that fluoride, lanthanum, and aluminum were the main FTLA components on the dentin surface after 6,000-stroke abrasion. To measure fluoride release from the FTLA components, a slurry was enclosed in a cellulose tube and suspended in deionized water at 37 degrees C. After fluoride was dialyzed against deionized water, a high concentration of fluoride was found to be released from FTLA the components, indicating FTLA treatment's prominent durability. These results suggested that FTLA treatment has a superior resistance against toothbrush abrasion and a high fluoride-releasing performance. These characteristics lend much weight to showing that the FTLA method is an effective and durable medicament for dentin hypersensitivity.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
449857
Lanthanum(III) fluoride, anhydrous, powder, 99.99% trace metals basis
F3La