EMAIL THIS PAGE TO A FRIEND
The European journal of prosthodontics and restorative dentistry
The effect of a range of disinfectants on the dimensional accuracy of some impression materials.
PMID 15691188
Abstract
In this study the dimensional accuracy of two model materials; dental stone and plaster of Paris, reproduced from three commonly used impression materials; alginate, polyether and addition-cured silicone, retained by their adhesives in acrylic resin trays and exposed to four disinfectant solutions was evaluated. Ninety casts were used to investigate the effect of the four disinfectants on the dimensional accuracy of alginate, polyether and addition-cured silicone impression material. For each impression material 30 impressions were taken, half were poured in dental stone and half in plaster of Paris. The disinfectants used were Dimenol, Perform-ID, MD-520, and Haz-tabs. Measurements were carried out using a High Precision Reflex Microscope. For the alginate impressions only those disinfected by 5-minute immersion in Haz-tabs solution and in full-strength MD 520 were not adversely affected by the disinfection treatment. All polyether impressions subjected to immersion disinfection exhibited a clinically acceptable expansion. Disinfected addition-cured silicone impressions produced very accurate stone casts. Those disinfected by spraying with fill-strength Dimenol produced casts that were very similar to those left as controls, but those treated by immersion disinfection exhibited negligible and clinically acceptable expansion. The results of the studied demonstrated that the various disinfection treatments had different effects on the impression materials. It is important that an appropriate disinfectant is used for each type of impression material.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
218928
Sodium dichloroisocyanurate, 96%
C3Cl2N3NaO3
35915
Sodium dichloroisocyanurate dihydrate, ≥98.0% (AT)
C3Cl2N3NaO3 · 2H2O