EMAIL THIS PAGE TO A FRIEND
The Journal of biological chemistry
Substrate enhances the sensitivity of type I protein kinase a to cAMP.
PMID 15691833
Abstract
The functional significance of the presence of two major (types I and II) isoforms of the cAMP-dependent protein kinase (PKA) is still enigmatic. The present study showed that peptide substrate enhanced the activation of PKA type I at low, physiologically relevant concentrations of cAMP through competitive displacement of the regulatory RI subunit. The effect was similar whether the substrate was a short peptide or the physiological 60-kDa protein tyrosine hydroxylase. In contrast, substrate failed to affect the cAMP-sensitivity of PKA type II. Size exclusion chromatography confirmed that substrate acted to physically enhance the dissociation of the RIalpha and Calpha subunits of PKA type I, but not the RIIalpha and Calpha subunits of PKA type II. Substrate availability can therefore fine-tune the activation of PKA type I by cAMP, but not PKA type II. The cAMP-dissociated RII and C subunits of PKA type II reassociated much faster than the PKA type I subunits in the presence of substrate peptide. This suggests that only PKA type II is able to rapidly reverse its activation after a burst of cAMP when exposed to high substrate concentration. We propose this as a possible reason why PKA type II is preferentially found in complexes with substrates undergoing rapid phosphorylation cycles.