EMAIL THIS PAGE TO A FRIEND
Molecular and Cellular Proteomics
Analysis of polyubiquitin conjugates reveals that the Rpn10 substrate receptor contributes to the turnover of multiple proteasome targets.
PMID 15699485