EMAIL THIS PAGE TO A FRIEND
Mycopathologia
The effects of the Penicillium mycotoxins citrinin, cyclopiazonic acid, ochratoxin A, patulin, penicillic acid, and roquefortine C on in vitro proliferation of porcine lymphocytes.
PMID 15702270
Abstract
The in vitro effect of each of the Penicillium mycotoxins citrinin (CIT), cyclopiazonic acid (CPA), ochratoxin A (OTA), patulin (PAT), penicillic acid (PIA) and roquefortine C (RQC) on mitogen induced lymphocyte proliferation was determined using purified lymphocytes from 6 piglets. Dose response curves for each mycotoxin were generated and the concentrations producing 50% inhibition of cell proliferation (IC(50)) were estimated. OTA and PAT were the most potent toxins with IC(50) of 1.3 and 1.2 micromol/l, respectively (0.52 and 0.18 mg/l, respectively). Based on molar concentrations, OTA was 15, 30, 40, and 65 times more potent as an inhibitor than PIA, CIT, CPA and RQC, respectively.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SML0406 Roquefortine C, ≥98% (HPLC), from Penicillium roqueforti
C22H23N5O2