EMAIL THIS PAGE TO A FRIEND
Organic letters
An expeditious synthesis of DPD and boron binding studies.
PMID 15704896
Abstract
A practical synthesis has been developed for DPD (4,5-dihydroxypentane-2,3-dione), an unstable small molecule that is proposed to be the source of universal signaling agents for quorum sensing in bacteria. The synthesis allows preparation of isotopically labeled DPD and ent-DPD as well as detailed studies of spontaneous binding to borate to give the unusual borate complex 6, the signal for marine bacteria such as Vibrio harveyi. [reaction: see text]