EMAIL THIS PAGE TO A FRIEND
Water research
Biomass characteristics in three sequencing batch reactors treating a wastewater containing synthetic organic chemicals.
PMID 15707644
Abstract
The physical and biochemical characteristics of the biomass in three lab-scale sequencing batch reactors (SBR) treating a synthetic wastewater at a 20-day target solids retention time (SRT) were investigated. The synthetic wastewater feed contained biogenic compounds and 22 organic priming compounds, chosen to represent a wide variety of chemical structures with different N, P and S functional groups. At a two-day hydraulic retention time (HRT), the oxidation-reduction potential (ORP) cycled between -100 (anoxic) and 100 mV (aerobic) in the anoxic/aerobic SBR, while it remained in a range of 126+/-18 and 249+/-18 mV in the aerobic sequencing batch biofilm reactor (SBBR) and the aerobic SBR reactor, respectively. A granular activated sludge with excellent settleability (SVI=98+/-31 L mg(-1)) developed only in the anoxic/aerobic SBR, compared to a bulky sludge with poor settling characteristics in the aerobic SBR and SBBR. While all reactors had very good COD removal (>90%) and displayed nitrification, substantial nitrogen removal (74%) was only achieved in the anoxic/aerobic SBR. During the entire operational period, benzoate, theophylline and 4-chlorophenol were completely removed in all reactors. In contrast, effluent 3-nitrobenzoate was recorded when its influent concentration was increased to 5 mg L(-1) and dropped only to below 1 mg L(-1) after 300 days of operation. The competent (active) biomass fractions for these compounds were between 0.04% and 5.52% of the total biomass inferred from substrate-specific microbial enumerations. The measured competent biomass fractions for 4-chlorophenol and 3-nitrobenzoate degradation were significantly lower than the influent COD fractions of these compounds. Correspondent to the highest competent biomass fraction for benzoate degradation among the test SOCs, benzoate oxidation could be quantified with an extant respirometric technique, with the highest specific oxygen uptake rate (SOUR(benzoate), 0.026 g O2 h(-1) g(-1) XCOD) in the anoxic/aerobic SBR. These combined results suggest that operating SBRs with alternative anoxic/aerobic cycles might facilitate the formation of granular sludge with good settleability, and retain comparable removal of nitrogen and synthetic organic compounds. Hence, the practice of anoxic/aerobic cycling should be considered in wastewater treatment systems whenever possible.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
185329
3-Nitrobenzoic acid, ReagentPlus®, 99%
C7H5NO4
72915
Sodium 3-nitrobenzoate, ≥95.0% (HPLC)
C7H4NNaO4