EMAIL THIS PAGE TO A FRIEND
Biochemistry
Isotope effects for deuterium transfer between substrate and coenzyme in adenosylcobalamin-dependent glutamate mutase.
PMID 15709782
Abstract
A key step in the mechanism of all adenosylcobalamin-dependent enzymes is the abstraction of a hydrogen atom from the substrate by a 5'-deoxyadenosyl radical generated by homolytic fission of the coenzyme cobalt-carbon bond. We have investigated the isotope effects associated with this process for glutamate mutase reacting with deuterated glutamate. The kinetics of deuterium incorporation into 5'-deoxyadenosine (5'-dA) during the reaction were followed by rapid chemical quench, using HPLC and electrospray mass spectrometry to analyze the 5'-dA formed. The kinetics of 5'-dA formation are biphasic, comprising a rapid phase k(app) = 37 +/- 3 s(-)(1) and a slower phase k(app) = 0.9 +/- 0.4 s(-)(1). The mass spectral data clearly show that the faster phase is associated with the formation of monodeuterated 5'-dA whereas the slower phase is associated with the incorporation of a second and then a third deuterium into 5'-dA. This observation implies that a large inverse equilibrium secondary isotope effect is associated with the formation of 5'-dA from adenosylcobalamin. The primary deuterium kinetic isotope effects on V and V/K for the formation of 5'-dA were determined from time-based and competition experiments. (D)V = 2.4 +/-0.4 whereas (D)(V/K) = 10 +/- 0.4, implying that an isotopically insensitive step is partially rate-determining. The additional data provided by these experiments cause us to revise our interpretation of earlier UV-visible stopped-flow kinetic measurements of AdoCbl homolysis obtained with deuterated substrates.
Related Materials