EMAIL THIS PAGE TO A FRIEND
The Veterinary record
Consequences of the withdrawal of dimetridazole on intestinal parasitism in ducks.
PMID 15717397
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
D4025
Dimetridazole
C5H7N3O2
31707
Dimetridazole, VETRANAL®, analytical standard
C5H7N3O2