EMAIL THIS PAGE TO A FRIEND
Chemical communications (Cambridge, England)
Ring expansion of sulfur substituted p-quinamines: regiospecific synthesis of 4-aminotropones.
PMID 15719098
Abstract
Synthesis of 4-aminotropones through a cyclization-ring expansion process occurs in a single step and with excellent yields from 4-amino-2,5-cyclohexadienones (p-quinamines) bearing a 4-sulfinyl or sulfonyl methyl group.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
252832
Tropone, 97%
C7H6O