EMAIL THIS PAGE TO A FRIEND
British Journal of Pharmacology
Validation of IKK beta as therapeutic target in airway inflammatory disease by adenoviral-mediated delivery of dominant-negative IKK beta to pulmonary epithelial cells.
PMID 15723090