EMAIL THIS PAGE TO A FRIEND
Dermatitis : contact, atopic, occupational, drug
Allergic contact dermatitis from cocamidopropyl betaine, cocamidoamine, 3-(dimethylamino)propylamine, and oleamidopropyl dimethylamine: co-reactions or cross-reactions?
PMID 15724349
Abstract
We present the case of a patient with facial dermatitis caused by sensitization to cocamidopropyl betaine. The patient also had positive patch-test reactions to cocamidoamine, 3-(dimethylamino)propylamine, and oleamidopropyl dimethylamine. The presence of 3-(dimethylamino)propylamine as an impurity in all of these substances can be hypothesized to explain these simultaneous reactions.
Related Materials