EMAIL THIS PAGE TO A FRIEND
Hinyokika kiyo. Acta urologica Japonica
[Combination chemotherapy with docetaxel and cisplatin in patients with hormone-refractory prostate cancer].
PMID 15732332
Abstract
We investigated the efficacy of docetaxel and cisplatin for hormone-refractory prostate cancer (HRPC). Thirteen patients with HRPC were treated with 30 mg/m2 docetaxel weekly for 3 weeks and 70 mg/m2 cisplatin on day 1. Treatment was repeated every 21 days. They received 2 cycles and were evaluated for the responses to serum prostate-specific antigen (PSA) and tumor size. Ten (77%) of the 13 patients showed a 50% or greater decrease in PSA with a median time to progression of 3 months. One of the 2 patients with measurable soft tissue disease showed a reduction in disease. No severe toxicity of this regimen was observed. Combination chemotherapy with docetaxel and cisplatin in patients with HRPC was well tolerated and efficatious with a significant decrease in serum PSA and measurable disease.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
D3881
Diethylstilbestrol dipropionate, analytical standard, for drug analysis
C24H28O4