EMAIL THIS PAGE TO A FRIEND
Journal of medicinal chemistry
SC-53116: the first selective agonist at the newly identified serotonin 5-HT4 receptor subtype.
PMID 1573641
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
PZ0146
SC-53116 hydrochloride hydrate, ≥98% (HPLC)
C16H22ClN3O2 · HCl · xH2O