EMAIL THIS PAGE TO A FRIEND
Journal of agricultural and food chemistry
Characterization and quantification of free and esterified 9- and 13-hydroxyoctadecadienoic acids (HODE) in barley, germinating barley, and finished malt.
PMID 15740040
Abstract
The analysis of (R)-9- and (S)-9-hydroxy-10E,12Z-octadecadienoic acid as well as (R)-13- and (S)-13-hydroxy-9Z,11E-octadecadienoic acid (HODE) as free acids, esterified in triacylglycerols (storage lipids), and esterified in polar lipids (phospholipids, glycolipids, etc.) in barley, germinating barley, and finished malt was performed using [13-(18)O(1)]-(S)-13-HODE isotope dilution assays with GC-MS and straight- and chiral-phase HPLC. 9- and 13- HODE occur approximately racemically in barley, indicating an autoxidation. The enantiomeric excesses increase to 78% S for free 9-HODE and to 58% S for free 13-HODE in germinating barley as a result of lipoxygenase-2 (LOX-2) catalysis, but free HODEs are at low concentration. More than 90% of HODEs in barley and malt are esterified. In the storage lipids of green malt 53 mg/kg 9-HODE and 147 mg/kg 13-HODE were detected. This ratio of 30:70 reflects the regioselectivity of the LOX-2 enzyme in malt. In the polar lipids 45 mg/kg 9-HODE and 44 mg/kg 13-HODE were characterized. The latter indicate a hitherto unknown 9-lipoxygenase activity with polar lipids as substrates. During kilning the contents of most HODEs decreased significantly due to chemical and enzymatic degradation, whereas polar-esterified (R)-13-HODE increased (43%) in the finished malt.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SML0503
9(S)-HODE, ≥98% (HPLC)
C18H32O3