EMAIL THIS PAGE TO A FRIEND
Biochemical pharmacology
Involvement of SULT1A3 in elevated sulfation of 4-hydroxypropranolol in Hep G2 cells pretreated with beta-naphthoflavone.
PMID 15748705
Abstract
Pretreatment of Hep G2 cells with beta-naphthoflavone (BNF 1-25microM) significantly increased cytosolic sulfation activities of 4-hydroxypropranolol (4-OH-PL) racemate. The profile was similar to those of sulfations towards dopamine and triiodothyronine in the same cytosolic fractions. Kinetic studies of 4-OH-PL sulfation in Hep G2 cytosolic fractions revealed that V(max) values increased but apparent K(m) values remained unchanged following the BNF pretreatment. Among five recombinant human SULT isoforms (SULT1A1, -1A3, -1B1, -1E1 and -2A1) examined, only SULT2A1 did not show 4-OH-PL sulfation activities under the conditions used. SULT1A3 and -1E1 exhibited an enantioselectivity of 4-OH-R-PL sulfation>4-OH-S-PL sulfation, which agreed with that of BNF-pretreated Hep G2 cells as well as of nontreated cells, whereas SULT1A1 and -1B1 showed a reversed enantioselectivity (R
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
00881
(±)-4-Hydroxypropranolol hydrochloride, analytical standard
C16H21NO3·HCl