EMAIL THIS PAGE TO A FRIEND
Reproductive toxicology (Elmsford, N.Y.)
Antiandrogenic effects in male rats perinatally exposed to a mixture of di(2-ethylhexyl) phthalate and di(2-ethylhexyl) adipate.
PMID 15749265
Abstract
Di(2-ethylhexyl) phthalate (DEHP) is a well-known testicular toxicant inducing adverse effects in androgen responsive tissues. Therefore, di(2-ethylhexyl) adipate (DEHA) is currently being evaluated as a potential substitute for DEHP. Similarities in structure and metabolism of DEHP and DEHA have led to the hypothesis that DEHA can modulate the effects of DEHP. Wistar rats were gavaged with either vehicle, DEHP (300 or 750mg/kg bw/day) or DEHP (750mg/kg bw/day) in combination with DEHA (400mg/kg bw/day) from gestation day (GD) 7 to postnatal day (PND) 17. Decreased anogenital distance (AGD) and retention of nipples in male offspring were found in all three exposed groups. Dosed males exhibited decreased weights of ventral prostate and m. levator ani/bulbocavernosus. Histopathological investigations revealed alterations in testis morphology in both juvenile and adult animals. The litter size was decreased and postnatal mortality was increased in the combination group only, which is likely a combined effect of DEHP and DEHA. However, no combination effect was seen with respect to antiandrogenic effects, as males receiving DEHP in combination with DEHA did not exhibit more pronounced effects in the reproductive system than males receiving DEHP alone.