EMAIL THIS PAGE TO A FRIEND
Free radical biology & medicine
Beryllium-stimulated reactive oxygen species and macrophage apoptosis.
PMID 15749389
Abstract
Beryllium (Be), the etiologic agent of chronic beryllium disease, is a toxic metal that induces apoptosis in human alveolar macrophages. We tested the hypothesis that Be stimulates the formation of reactive oxygen species (ROS) which plays a role in Be-induced macrophage apoptosis. Mouse macrophages were exposed to 100 microM BeSO4 in the absence and presence of the catalytic antioxidant MnTBAP (100 microM). Apoptosis was measured as the percentage of TUNEL+ and caspase-8+ cells. ROS production was measured by flow cytometry using the fluorescence probes, dihydroethidine (DHE) and dichlorofluorescein diacetate (DCFH-DA). Be-exposed macrophages had increased TUNEL+ cells (15+/-1% versus controls 1+/-0.2%, P<0.05) and increased caspase-8+ cells (18.7+/-2% versus controls 1.8+/-0.4%, P<0.05). Be-induced caspase-8 activation, and a 4-fold increase in ROS formation, was ameliorated by exposure to MnTBAP. Hydrogen peroxide (30 microM) exposure potentiated Be-induced caspase-8 activation, and was also attenuated by MnTBAP. Our data are the first to demonstrate that Be stimulates macrophage ROS formation which plays an important role in Be-induced macrophage apoptosis.
Related Materials