EMAIL THIS PAGE TO A FRIEND
Fitoterapia
Antibacterial and antifungal activity of pinosylvin, a constituent of pine.
PMID 15752644
Abstract
The antibacterial and antifungal activities of pinosylvin (3,5-dihydroxy-trans-stilbene), a constituent of pine, were studied and compared with those of resveratrol (3,5,4'-trihydroxy-trans-stilbene). Pinosylvin exhibited more potent growth inhibitory activity against Candida albicans and Saccharomyces cerevisiae.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
56297
Pinosylvin, ≥97.0% (HPLC)
C14H12O2