EMAIL THIS PAGE TO A FRIEND
Medicina clinica
[Dichloropropane poisoning: report of 2 cases].
PMID 15755402
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
02577
1,2-Dichloropropane, analytical standard
C3H6Cl2
82270
1,2-Dichloropropane, 99%
C3H6Cl2