EMAIL THIS PAGE TO A FRIEND
Chemical communications (Cambridge, England)
Nickel catalysed asymmetric cycloisomerisation of diethyl diallylmalonate.
PMID 15756334
Abstract
Cationic nickel catalysts with monodentate phosphoramidites and Wilke's azaphospholene as ligands are highly regio- and enantioselective catalysts for the cycloisomerisation of diethyl diallylmalonate.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
283479
Diethyl diallylmalonate, 98%
C13H20O4