EMAIL THIS PAGE TO A FRIEND
Biochemical and biophysical research communications
A genetically engineered purpurin/retinol-binding protein hybrid that binds to transthyretin.
PMID 1575761
Abstract
A mini-gene encoding rat retinol-binding protein (RBP) and a cDNA encoding chicken purpurin were separately transfected into HeLa cells. In contrast to RBP, expressed purpurin did not bind to transthyretin (TTR). A purpurin/RBP hybrid protein was constructed by substituting the cDNA sequence encoding the N-terminal 29 amino acids of purpurin for the corresponding part of RBP. The expressed hybrid molecule bound to the TTR-Sepharose. These results demonstrate that purpurin does not bind to TTR, that a functional purpurin/RBP hybrid can be constructed, and that the N-terminal coil of RBP is not required for TTR binding.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
82631
Purpurin, for spectrophotometric det. (of B, Pb)
C14H8O5
229148
Purpurin, Dye content 90 %
C14H8O5