EMAIL THIS PAGE TO A FRIEND
The Journal of organic chemistry
Ionic liquid-immobilized quinuclidine-catalyzed Morita-Baylis-Hillman reactions.
PMID 15760226
Abstract
[reaction: see text] The ionic liquid-bound quinuclidine catalyzed Baylis-Hillman reactions were investigated. The IL-supported catalyst showed equally good catalytic activity as compared with its nonimmobilized counterpart. The corresponding Baylis-Hillman adducts were obtained in moderate to high yields in all the cases tested. The IL-supported quinuclidine can be readily recovered and reused six times without significant loss of catalytic activity.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
197602
Quinuclidine, 97%
C7H13N