EMAIL THIS PAGE TO A FRIEND
Life sciences
Human endothelial nitric oxide synthase gene delivery protects against cardiac remodeling and reduces oxidative stress after myocardial infarction.
PMID 15763077
Abstract
Nitric oxide (NO) has been shown to play a key role in the regulation of cardiac hypertrophy and fibrosis in response to myocardial ischemia in part by antagonizing the action of angiotensin II (Ang II). In this study, we investigated the potential protective role of human endothelial nitric oxide synthase (eNOS) in left ventricular (LV) remodeling after myocardial infarction (MI) by a somatic gene transfer approach. Male Wistar rats underwent coronary artery ligation to induce MI. One week after surgery, adenovirus encoding the human eNOS or luciferase gene under the control of the CMV promoter/enhancer was injected into rats via the tail vein, and animals were sacrificed at 1 and 5 weeks after gene transfer. Successful gene transfer was evaluated based on increased levels of NO and cGMP in the heart, measured at one week after eNOS gene delivery. Six weeks after MI, the LV end-diastolic pressure, heart weight, LV axis length and cardiomyocyte size were markedly increased compared to the Sham group, while eNOS gene delivery significantly reduced these parameters. Rats receiving control virus developed considerably more fibrotic lesions identified by Sirius Red staining and collagen I immunostaining compared to Sham rats, and eNOS gene delivery significantly reduced collagen accumulation. eNOS gene transfer also reduced TUNEL-positive apoptotic cells. The cardioprotective effect of NO was accompanied by reduced NADH and NADPH oxidase activities and superoxide formation, TGF-beta1 and p27 levels, JNK activation, NF-kappa B nuclear translocation, and caspase-3 activity. This study shows that NO may play an important role in attenuating cardiac remodeling and apoptosis after myocardial infarction via suppression of oxidative stress-mediated signaling pathways.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
195251
Direct Red 81, Dye content 50 %
C29H19N5Na2O8S2