EMAIL THIS PAGE TO A FRIEND
Steroids
Steroids' transformations in Penicillium notatum culture.
PMID 15763598
Abstract
The application of Penicillium notatum genus for biotransformations of steroids has been investigated. The reactions observed include insertion of an oxygen atom into D-ring of steroids, 15alpha-hydroxylation of 17alpha-methyl testosterone derivatives, ester bond hydrolysis, and degradation of a testosterone derivatives side chain. Microbial production of testolactones, the biologically active compounds, was also achieved using this strain in up to 98% yield.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
E1001
Ethisterone, ≥99% (HPLC)
C21H28O2
46272
Ethisterone, VETRANAL, analytical standard
C21H28O2