EMAIL THIS PAGE TO A FRIEND
Archives of physiology and biochemistry
Effects of an alpha2-adrenoceptor agonist in nasal mucosa.
PMID 15764068
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
B162
B-HT 920 dihydrochloride, solid, ≥98% (HPLC)
C10H15N3S · 2HCl