EMAIL THIS PAGE TO A FRIEND
Communications in agricultural and applied biological sciences
Weed control and persistence of two oxyfluorfen formulations in olive groves under non tillage conditions.
PMID 15765564
Abstract
To obtain profitable yields in olive groves, residual preemergence herbicides are applied in October or November before the winter rains, and before the winter annual weeds germinate. Simazine, one of the herbicides most used for weed control in olive groves, has recently been banned. Oxyfluorfen is presented as a good alternative to simazine in olive fields. Experiments were carried out in 2002 and 2003 to evaluate the behaviour of two oxyfluorfen formulations, 2XL and G4F, at 480 g a.i. ha(-1) for three different soil management systems with three replications (1. bare soil; 2. bare soil and grassed buffer strips, chemically controlled and 3. bare soil and grassed buffer strips with controlled mowing; under non tillage conditions in all three cases). The most important species that survived 2XL and G4F treatments was Sagina apetala ARD. Oxyfluorfen residues were evaluated throughout 158 days after the applications. Three soil samples from each plot were collected, mixed and air dried. The herbicide extractions were made with methanol and the residues were analyzed by HPLC. We found no differences between the two formulations, but results showed that recoveries of oxyfluorfen were higher in plots with chemically controlled buffer grassed strips than in the other soil management types.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
35031
Oxyfluorfen, PESTANAL®, analytical standard
C15H11ClF3NO4