EMAIL THIS PAGE TO A FRIEND
The Journal of biological chemistry
Novel small molecule inhibitors of 3-phosphoinositide-dependent kinase-1.
PMID 15772071
Abstract
The phosphoinositide 3-kinase/3-phosphoinositide-dependent kinase 1 (PDK1)/Akt signaling pathway plays a key role in cancer cell growth, survival, and tumor angiogenesis and represents a promising target for anticancer drugs. Here, we describe three potent PDK1 inhibitors, BX-795, BX-912, and BX-320 (IC(50) = 11-30 nm) and their initial biological characterization. The inhibitors blocked PDK1/Akt signaling in tumor cells and inhibited the anchorage-dependent growth of a variety of tumor cell lines in culture or induced apoptosis. A number of cancer cell lines with elevated Akt activity were >30-fold more sensitive to growth inhibition by PDK1 inhibitors in soft agar than on tissue culture plastic, consistent with the cell survival function of the PDK1/Akt signaling pathway, which is particularly important for unattached cells. BX-320 inhibited the growth of LOX melanoma tumors in the lungs of nude mice after injection of tumor cells into the tail vein. The effect of BX-320 on cancer cell growth in vitro and in vivo indicates that PDK1 inhibitors may have clinical utility as anticancer agents.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SML0694
BX-795 hydrochloride, ≥98% (HPLC)
C23H26IN7O2S · xHCl
SML1086 BX-912, ≥95% (HPLC)
C20H23BrN8O