EMAIL THIS PAGE TO A FRIEND
Cancer Research
Hair follicle-derived blood vessels vascularize tumors in skin and are inhibited by Doxorubicin.
PMID 15781648
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
CBL1337 Anti-PECAM-1 Antibody, clone 390, clone 390, Chemicon®, from rat