EMAIL THIS PAGE TO A FRIEND
Antimicrobial agents and chemotherapy
Accumulation and oriented transport of ampicillin in Caco-2 cells from its pivaloyloxymethylester prodrug, pivampicillin.
PMID 15793098
Abstract
Pivampicillin (PIVA), an acyloxymethylester of ampicillin, is thought to enhance the oral bioavailability of ampicillin because of its greater lipophilicity compared to that of ampicillin. The fate of PIVA in intestinal cells and the exact location of its conversion into ampicillin have, however, never been unambiguously established. Polarized Caco-2 cells have been used to examine the handling of PIVA and the release of ampicillin from PIVA by the intestinal epithelium. Experiments were limited to 3 h. Cells incubated with PIVA (apical pole) showed a fast accumulation of ampicillin and transport toward the basolateral medium, whereas PIVA itself was only poorly accumulated and transported. Cells incubated with free ampicillin accumulated and transported only minimal amounts of this drug. Release of ampicillin from cells incubated with PIVA was unaffected by PEPT1 and OCTN2 inhibitors but was sharply decreased after ATP depletion or addition of bis(4-nitrophenyl)-phosphate (BNPP; an esterase inhibitor). PIVA incubated with Caco-2 lysates released free ampicillin, and this release was inhibited by BNPP. Efflux studies showed that the ampicillin that accumulated in cells after incubation with PIVA was preferentially transported out of the cells through the basolateral pole. This efflux was decreased by multidrug resistance-associated protein (MRP) inhibitors (probenecid, MK-571) and by ATP depletion. A phthalimidomethylester of ampicillin that resists cellular esterases failed to cause any significant release (cell lysate) or transport (polarized Caco-2 cells) of ampicillin. These results show that when PIVA is given to Caco-2 cells from their apical pole, ampicillin is released intracellularly and that ampicillin is thereafter preferentially effluxed into the basolateral medium through an MRP-like transporter.