EMAIL THIS PAGE TO A FRIEND
Chemical communications (Cambridge, England)
Dynamic synthesis of a macrocycle containing a porphyrin and an electron donor.
PMID 15795761
Abstract
New macrocycles incorporating a porphyrin and a [small pi] electron-rich aromatic were prepared from a dynamic disulfide library. The outcome could be influenced by use of templates.