EMAIL THIS PAGE TO A FRIEND
Presse medicale (Paris, France : 1983)
[Oesophageal ulcer associated with the use of bacampicillin].
PMID 15798551
Abstract
Many drugs may provoke oesophageal disorders. In France, doxycyclin, potassium chloride non-steroidal anti-inflammatories are the most frequent agents incriminated. A 27 year-old man consulted for dysphagia and odynophagia due to oesophagitis following administration of bacampicillin. Several drugs can provoke oesophageal disorders including ulcerations. Predisposing circumstances exist. Ingestion of the drugs with a sufficient quantity of water (60 to 100 ml), whilst standing or sitting should be able to limit this type of event.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
B0070000 Bacampicillin hydrochloride, European Pharmacopoeia (EP) Reference Standard
C21H27N3O7S · HCl