EMAIL THIS PAGE TO A FRIEND
Bioorganic & medicinal chemistry
Synthesis and pharmacological evaluation of benzamide derivatives as selective 5-HT(4) receptor agonists.
PMID 15809165
Abstract
It is thought that selective 5-HT(4) receptor agonists-such as 4-amino-5-chloro-2-methoxy-N-[1-(6-oxo-6-phenylhexyl)piperidin-4ylmethyl]benzamide (2)-have the ability to enhance both upper and lower gastrointestinal motility without any significant adverse effects. Modification of 2 was performed. Variation of the piperidin-4ylmethyl moiety of 2 led to a decrease in the binding affinity for the 5-HT(4) receptor. Following conversion of the carbonyl group on the benzoyl part to a hydroxyl or sulfoxide group, the binding affinity for the 5-HT(4) receptor was retained although the effect on defecation was reduced. Many of the 4-amino-5-chloro-2-methoxy-N-(piperidin-4ylmethyl)benzamides that had a ether or sulfide moiety in the side-chain part at the 1-position of the piperidine exhibited high affinity for the 5-HT(4) receptor. Among these, phenylthio 41c and benzylthio derivative 44 were selective 5-HT(4) receptor agonists, and had a similar effect on defecation to compound 2.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
143200
Ethyl 1-benzyl-3-oxo-4-piperidinecarboxylate hydrochloride, technical grade
C15H19NO3 · HCl