EMAIL THIS PAGE TO A FRIEND
Zhong yao cai = Zhongyaocai = Journal of Chinese medicinal materials
[Anti-tumor effects of ampelopsin on human lung cancer GLC-82 implanted in nude mice].
PMID 15810594
Abstract
To investigate the effects of ampelopsin on the growth of human lung cancer cell line GLC-82 in nude mice. GLC-82 cells were inoculated into the armpit of nude mice to establish lung cancer model. Then the BALB/C nude mice 6 with xenograft tumor were randomized into 6 groups. Ampelopsin was administered at 3 dosages by intraperitoneal injection daily. The anti-tumor effect of ampelopsin was evaluated on tumor volume, relative tumor volume, tumor weight, relative tumor proliferative rale, and tumor growth curve. At the 250 mg/kg dose of ampelopsin, the growth inhibition of the transplant tumor was 35.5% (P < 0.01) and 37.1% (P < 0.01), and the relative tumor proliferative rate was 55.24% (P < 0.01) and 57.71% (P < 0.05), respectively in two independent experiments by compared with the normal saline control. Ampelopsin appears to be a potent anti-tumor agent that was firstly discovered in the transplant human lung cancer cell line GLC-82 in nude mice in vivo.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SML0295
Dihydromyricetin, ≥98% (HPLC)
C15H12O8