EMAIL THIS PAGE TO A FRIEND
Chemosphere
Biosorption of water-soluble dyes on magnetically modified Saccharomyces cerevisiae subsp. uvarum cells.
PMID 15811411
Abstract
Brewer's yeast (bottom yeast, Saccharomyces cerevisiae subsp. uvarum) cells were magnetically modified using water based magnetic fluid stabilized with perchloric acid. Magnetically modified yeast cells efficiently adsorbed various water soluble dyes. The dyes adsorption can be described by the Langmuir adsorption model. The maximum adsorption capacity of the magnetic cells differed substantially for individual dyes; the highest value was found for aniline blue (approx. 220 mg per g of dried magnetic adsorbent).
Related Materials